ENA238711608 Enamine

[1-(3-methylbutyl)-1H-1,3-benzodiazol-2-yl]methanol

  • MDL number MFCD02630594
  • Cas Number : 380543-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48616
Availability details for
Quantity
  • Availability
  • Add to Cart