ENA238711610 Enamine

propan-2-yl 2-(2-methyl-1H-1,3-benzodiazol-1-yl)acetate

  • MDL number MFCD02729777
  • Cas Number : 933944-55-9
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48620
Availability details for
Quantity
  • Availability
  • Add to Cart