ENA238711619 Enamine

4-(3-bromophenyl)oxane-4-carbonitrile

  • MDL number MFCD08277238
  • Cas Number : 245439-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48630
Availability details for
Quantity
  • Availability
  • Add to Cart