ENA238711624 Enamine

4-(3,4-dichlorophenyl)butanoic acid

  • MDL number MFCD02167688
  • Cas Number : 25157-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-72
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48746
Availability details for
Quantity
  • Availability
  • Add to Cart