ENA238711630 Enamine

[(2-ethoxyphenyl)methyl](methyl)amine hydrochloride

  • MDL number MFCD07106802
  • Cas Number : 104294-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49074
Availability details for
Quantity
  • Availability
  • Add to Cart