ENA238711631 Enamine

2-(3-fluorophenyl)-1H-1,3-benzodiazol-5-amine dihydrochloride

  • MDL number MFCD12197235
  • Cas Number : 1193389-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49075
Availability details for
Quantity
  • Availability
  • Add to Cart