ENA238711633 Enamine

1-(2-bromoethoxy)-2-methoxybenzene

  • MDL number MFCD00017892
  • Cas Number : 4463-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39...41
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49077

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-49077
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart