ENA238711634 Enamine

2-amino-N-(3-ethynylphenyl)acetamide hydrochloride

  • MDL number MFCD12197369
  • Cas Number : 1193389-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49078

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49078
Availability details for
Quantity
  • Availability
  • Add to Cart