ENA238711635 Enamine

4-methoxy-2-(piperazin-1-yl)pyrimidine dihydrochloride

  • MDL number MFCD12197370
  • Cas Number : 1193389-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49079
Availability details for
Quantity
  • Availability
  • Add to Cart