ENA238711652 Enamine

2-{[(2-methoxyphenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD05863675
  • Cas Number : 109926-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49098

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49098
Availability details for
Quantity
  • Availability
  • Add to Cart