ENA238711654 Enamine

[2-(dimethylamino)ethyl][(pyridin-3-yl)methyl]amine

  • MDL number MFCD03856646
  • Cas Number : 136469-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49100
Availability details for
Quantity
  • Availability
  • Add to Cart