ENA238711661 Enamine

1-acetamidocyclohexane-1-carboxylic acid

  • MDL number MFCD09933315
  • Cas Number : 4854-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49111

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49111
Availability details for
Quantity
  • Availability
  • Add to Cart