ENA238711666 Enamine

1-ethyl-4-(piperidin-4-yl)piperazine trihydrochloride

  • MDL number MFCD12197083
  • Cas Number : 1181457-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 324...326
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44174

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44174
Availability details for
Quantity
  • Availability
  • Add to Cart