ENA238711676 Enamine

4-methoxybutan-2-one

  • MDL number MFCD08704277
  • Cas Number : 6975-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49430
Availability details for
Quantity
  • Availability
  • Add to Cart