ENA238711677 Enamine

2-(2-iodobenzamido)thiophene-3-carboxylic acid

  • MDL number MFCD09046382
  • Cas Number : 926259-86-1
  • Molecular Weight : 373.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49483
Availability details for
Quantity
  • Availability
  • Add to Cart