ENA238711684 Enamine

4-(pyridin-3-ylmethoxy)phenol

  • MDL number MFCD11181724
  • Cas Number : 1094283-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49491

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49491
Availability details for
Quantity
  • Availability
  • Add to Cart