ENA238711696 Enamine

2-bromo-6-(4-fluorophenyl)imidazo[2,1-b][1,3,4]thiadiazole

  • MDL number MFCD11209935
  • Cas Number : 1094275-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49508
Availability details for
Quantity
  • Availability
  • Add to Cart