ENA238711701 Enamine

2-bromo-4-fluorobenzohydrazide

  • MDL number MFCD09727562
  • Cas Number : 935657-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49522

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49522
Availability details for
Quantity
  • Availability
  • Add to Cart