ENA238711704 Enamine

4-(furan-2-carbonyl)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

  • MDL number MFCD06660852
  • Cas Number : 926258-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49527
Availability details for
Quantity
  • Availability
  • Add to Cart