ENA238711706 Enamine

4-(3,5-dimethyl-1H-pyrazol-1-yl)-3-fluoroaniline

  • MDL number MFCD08699597
  • Cas Number : 1006468-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49529
Availability details for
Quantity
  • Availability
  • Add to Cart