ENA238711709 Enamine

2-(sulfanylmethyl)-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD09817354
  • Cas Number : 1016696-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49535
Availability details for
Quantity
  • Availability
  • Add to Cart