ENA238711712 Enamine

1-[(3,4-difluorophenyl)methyl]piperidin-4-amine

  • MDL number MFCD11048016
  • Cas Number : 160358-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49538
Availability details for
Quantity
  • Availability
  • Add to Cart