ENA238711720 Enamine

(2-bromo-5-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD11045216
  • Cas Number : 1170783-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44181

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44181
Availability details for
Quantity
  • Availability
  • Add to Cart