ENA238711724 Enamine

3-methanesulfonylpropan-1-amine hydrochloride

  • MDL number MFCD09744144
  • Cas Number : 157825-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49565

Safety Information

signalword Warning
safety_statement 7-16-36/37-45
msds https://www.enaminestore.com/catalog/EN300-49565
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart