ENA238711725 Enamine

2,5,7-trimethylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD12197249
  • Cas Number : 1158269-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226...228
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49568

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49568
Availability details for
Quantity
  • Availability
  • Add to Cart