ENA238711728 Enamine

(naphthalen-1-yl)(thiophen-2-yl)methanone

  • MDL number MFCD05149050
  • Cas Number : 56662-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49571

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49571
Availability details for
Quantity
  • Availability
  • Add to Cart