ENA238711731 Enamine

3-methyl-5-(4-methyl-3-nitrophenyl)-1,2,4-oxadiazole

  • MDL number MFCD12197250
  • Cas Number : 1193389-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49574
Availability details for
Quantity
  • Availability
  • Add to Cart