ENA238711732 Enamine

3-(1-benzofuran-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD09934965
  • Cas Number : 1107061-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49577
Availability details for
Quantity
  • Availability
  • Add to Cart