ENA238711734 Enamine

2-(azepan-1-yl)-N'-hydroxyethanimidamide

  • MDL number MFCD09043737
  • Cas Number : 98487-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49581

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49581
Availability details for
Quantity
  • Availability
  • Add to Cart