ENA238711738 Enamine

N-(pentan-3-yl)cyclopropanamine hydrochloride

  • MDL number MFCD12197251
  • Cas Number : 1193389-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49585

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-49585
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart