ENA238711747 Enamine

2-(ethylsulfanyl)aniline

  • MDL number MFCD08691983
  • Cas Number : 13920-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49598
Availability details for
Quantity
  • Availability
  • Add to Cart