ENA238711751 Enamine

N-butyl-4-(morpholin-4-yl)aniline

  • MDL number MFCD11161751
  • Cas Number : 1038225-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44183
Availability details for
Quantity
  • Availability
  • Add to Cart