ENA238711753 Enamine

[2-(cyclopropylmethoxy)phenyl]boronic acid

  • MDL number MFCD04039015
  • Cas Number : 1050510-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49611
Availability details for
Quantity
  • Availability
  • Add to Cart