ENA238711768 Enamine

4-amino-N-[4-chloro-3-(trifluoromethyl)phenyl]benzene-1-sulfonamide

  • MDL number MFCD12912775
  • Cas Number : 157887-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49628

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49628
Availability details for
Quantity
  • Availability
  • Add to Cart