ENA238711772 Enamine

4-amino-N-(2,4-dichlorophenyl)-2-methoxybenzene-1-sulfonamide

  • MDL number MFCD12912778
  • Cas Number : 1094926-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49634
Availability details for
Quantity
  • Availability
  • Add to Cart