ENA238711773 Enamine

4-amino-N-[(4-chlorophenyl)methyl]benzene-1-sulfonamide

  • MDL number MFCD08164713
  • Cas Number : 928001-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49635
Availability details for
Quantity
  • Availability
  • Add to Cart