ENA238711778 Enamine

6-(methylamino)pyridine-3-carboxylic acid

  • MDL number MFCD11132022
  • Cas Number : 262296-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49645
Availability details for
Quantity
  • Availability
  • Add to Cart