ENA238711782 Enamine

2-amino-N,N-dimethyl-2-phenylacetamide hydrochloride

  • MDL number MFCD12197255
  • Cas Number : 1214036-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-49650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49650
Availability details for
Quantity
  • Availability
  • Add to Cart