ENA238711784 Enamine

1-[2-(diethylamino)ethyl]-1H-pyrazol-4-amine

  • MDL number MFCD09965619
  • Cas Number : 1152841-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49654

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-49654
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart