ENA238711786 Enamine

ethyl 4-(chlorosulfonyl)butanoate

  • MDL number MFCD10011063
  • Cas Number : 319452-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49656
Availability details for
Quantity
  • Availability
  • Add to Cart