ENA238711788 Enamine

2-(4-aminophenyl)-N-ethyl-N-phenylacetamide

  • MDL number MFCD09730031
  • Cas Number : 953741-70-3
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49658
Availability details for
Quantity
  • Availability
  • Add to Cart