ENA238711793 Enamine

5-methyl-4-(3-nitrophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD11133113
  • Cas Number : 16942-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49688
Availability details for
Quantity
  • Availability
  • Add to Cart