ENA238711796 Enamine

N-[(4-bromophenyl)methyl]-2,4-difluoroaniline hydrochloride

  • MDL number MFCD12197256
  • Cas Number : 1193390-03-2
  • Molecular Weight : 334.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49692
Availability details for
Quantity
  • Availability
  • Add to Cart