ENA238711798 Enamine

2-(2,5-dioxo-4-propylimidazolidin-1-yl)acetic acid

  • MDL number MFCD09816023
  • Cas Number : 1104234-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49694
Availability details for
Quantity
  • Availability
  • Add to Cart