ENA238711807 Enamine

1-(3-fluoro-4-phenoxyphenyl)ethan-1-amine

  • MDL number MFCD11137327
  • Cas Number : 1019474-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49703
Availability details for
Quantity
  • Availability
  • Add to Cart