ENA238711810 Enamine

4-chloro-6,7,8-trifluoroquinoline-3-carbonitrile

  • MDL number MFCD09947327
  • Cas Number : 947339-99-3
  • Molecular Weight : 242.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-49710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49710
Availability details for
Quantity
  • Availability
  • Add to Cart