ENA238711813 Enamine

4-[(2-phenylethyl)amino]butanoic acid hydrochloride

  • MDL number MFCD12197259
  • Cas Number : 57054-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49713
Availability details for
Quantity
  • Availability
  • Add to Cart