ENA238711814 Enamine

3-methyl-5-(3-methyl-4-nitrophenyl)-1,2,4-oxadiazole

  • MDL number MFCD12197260
  • Cas Number : 1193389-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49715

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49715
Availability details for
Quantity
  • Availability
  • Add to Cart