ENA238711821 Enamine

N-[1-(2-cyanoethyl)-3-methyl-1H-pyrazol-5-yl]acetamide

  • MDL number MFCD12197086
  • Cas Number : 1181457-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44196

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44196
Availability details for
Quantity
  • Availability
  • Add to Cart