ENA238711822 Enamine

1-methyl-5-propanoyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD10694250
  • Cas Number : 1147762-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44198
Availability details for
Quantity
  • Availability
  • Add to Cart